Assay Details
| Gene Symbol: utp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199998.1 | NP_956292.1 | 1 - 2 | 187 | 108 | |
| GenBank mRNA | BC049053.1 | - | 1 - 2 | 187 | 108 | |
More Information
Set Membership: |
|

utp11
Genomic Map