Assay Details
| Gene Symbol: plcd1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200068.1 | NP_956362.1 | 4 - 5 | 715 | 147 | |
| XM_005163356.4 | XP_005163413.1 | 4 - 5 | 729 | 147 | ||
| GenBank mRNA | BC051157.1 | - | 4 - 5 | 715 | 147 | |
More Information
Set Membership: |
|

plcd1b
Genomic Map