Assay Details
| Gene Symbol: washc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200183.1 | NP_956477.1 | 22 - 23 | 3271 | 66 | |
| GenBank mRNA | BC045490.1 | - | 22 - 23 | 3271 | 66 | |
More Information
Set Membership: |
|

washc5
Genomic Map