Assay Details
| Gene Symbol: washc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200183.1 | NP_956477.1 | 285 | 87 | ||
| GenBank mRNA | BC045490.1 | - | 285 | 87 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

washc5| Gene Symbol: washc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200183.1 | NP_956477.1 | 285 | 87 | ||
| GenBank mRNA | BC045490.1 | - | 285 | 87 | ||
Set Membership: |
|