Assay Details
| Gene Symbol: hpda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201167.1 | NP_957461.1 | 12 - 13 | 1003 | 74 | |
| XM_005171729.3 | XP_005171786.1 | 12 - 13 | 1039 | 74 | ||
| GenBank mRNA | AY398361.1 | - | 11 - 12 | 967 | 74 | |
| BC046075.1 | - | 12 - 13 | 1003 | 74 | ||
More Information
Set Membership: |
|

hpda
Genomic Map