Assay Details
| Gene Symbol: chrna7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201219.2 | NP_957513.1 | 8 - 9 | 982 | 135 | |
| GenBank mRNA | AY247962.1 | - | 8 - 9 | 985 | 135 | |
| BC162599.1 | - | 8 - 9 | 970 | 135 | ||
| BC162602.1 | - | 8 - 9 | 970 | 135 | ||
More Information
Set Membership: |
|

chrna7a
Genomic Map