Assay Details
| Gene Symbol: fbxw11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201310.1 | NP_958467.1 | 11 - 12 | 1478 | 68 | |
| XM_005173069.4 | XP_005173126.1 | 12 - 13 | 1617 | 68 | ||
| GenBank mRNA | BC045356.1 | - | 11 - 12 | 1478 | 68 | |
More Information
Set Membership: |
|

fbxw11a
Genomic Map