Assay Details
| Gene Symbol: fbxw11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201310.1 | NP_958467.1 | 1 - 2 | 97 | 121 | |
| GenBank mRNA | BC045356.1 | - | 1 - 2 | 97 | 121 | |
More Information
Set Membership: |
|

fbxw11a
Genomic Map