Assay Details
| Gene Symbol: abcf2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201315.2 | NP_958472.2 | 11 - 12 | 1486 | 67 | |
| GenBank mRNA | BC047181.1 | - | 11 - 12 | 1461 | 67 | |
| BC066505.1 | - | 11 - 12 | 1390 | 67 | ||
| BC165771.1 | - | 10 - 11 | 1365 | 67 | ||
| BI709967.1 | - | 3 - 4 | 221 | 67 | ||
More Information
Set Membership: |
|

abcf2a
Genomic Map