Assay Details
| Gene Symbol: abcf2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201315.2 | NP_958472.2 | 6 - 7 | 970 | 77 | |
| GenBank mRNA | BC047181.1 | - | 6 - 7 | 945 | 77 | |
| BC066505.1 | - | 6 - 7 | 874 | 77 | ||
| BC165771.1 | - | 5 - 6 | 849 | 77 | ||
| CT713297.2 | - | 2 - 3 | 208 | 77 | ||
| EH577748.1 | - | 2 - 3 | 179 | 77 | ||
More Information
Set Membership: |
|

abcf2a
Genomic Map