Assay Details
| Gene Symbol: arcn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201459.1 | NP_958867.1 | 4 - 5 | 810 | 80 | |
| GenBank mRNA | BC045318.1 | - | 4 - 5 | 810 | 80 | |
| BC165628.1 | - | 3 - 4 | 675 | 80 | ||
More Information
Set Membership: |
|

arcn1a
Genomic Map