Assay Details
| Gene Symbol: lrrc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205539.1 | NP_991102.1 | 3 - 4 | 1107 | 72 | |
| XM_009300345.3 | XP_009298620.1 | 3 - 4 | 1245 | 72 | ||
| GenBank mRNA | BC065605.1 | - | 3 - 4 | 1107 | 72 | |
| CU468800.1 | - | 2 - 3 | 893 | 72 | ||
More Information
Set Membership: |
|

lrrc17
Genomic Map