Assay Details
| Gene Symbol: akt2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212815.2 | NP_997980.1 | 3 - 4 | 351 | 134 | |
| XM_021479143.1 | XP_021334818.1 | 434 | 134 | |||
| XM_021479144.1 | XP_021334819.1 | 442 | 134 | |||
| GenBank mRNA | BC046892.1 | - | 3 - 4 | 352 | 134 | |
| BC068106.1 | - | 3 - 4 | 319 | 134 | ||
| BC165215.1 | - | 2 - 3 | 201 | 134 | ||
More Information
Set Membership: |
|

akt2l
Genomic Map