Assay Details
| Gene Symbol: rab42a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213042.1 | NP_998207.1 | 1 - 2 | 243 | 60 | |
| GenBank mRNA | BC060898.1 | - | 1 - 2 | 243 | 60 | |
More Information
Set Membership: |
|

rab42a
Genomic Map