Assay Details
| Gene Symbol: tmem19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002098.1 | 818 | 72 | |||
| GenBank mRNA | BC071522.1 | - | 818 | 72 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tmem19| Gene Symbol: tmem19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002098.1 | 818 | 72 | |||
| GenBank mRNA | BC071522.1 | - | 818 | 72 | ||
Set Membership: |
|