Assay Details
| Gene Symbol: otud5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001327765.1 | NP_001314694.1 | 2 - 3 | 871 | 133 | |
| GenBank mRNA | BC078209.1 | - | 2 - 3 | 394 | 133 | |
More Information
Set Membership: |
|

otud5b
Genomic Map