Assay Details
| Gene Symbol: cdkn2aip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006085.1 | NP_001006085.1 | 3 - 4 | 427 | 88 | |
| XM_005169040.3 | XP_005169097.1 | 3 - 4 | 499 | 88 | ||
| XM_005169042.3 | XP_005169099.1 | 3 - 4 | 364 | 88 | ||
| GenBank mRNA | BC083234.1 | - | 3 - 4 | 427 | 88 | |
| BC164098.1 | - | 2 - 3 | 400 | 88 | ||
More Information
Set Membership: |
|

cdkn2aip
Genomic Map