Assay Details
| Gene Symbol: mettl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007336.1 | NP_001007337.1 | 6 - 7 | 736 | 64 | |
| XM_021478595.1 | XP_021334270.1 | 5 - 6 | 524 | 64 | ||
| GenBank mRNA | BC085548.1 | - | 6 - 7 | 736 | 64 | |
More Information
Set Membership: |
|

mettl8
Genomic Map