Assay Details
| Gene Symbol: fbxo44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008577.2 | NP_001008577.2 | 6 - 7 | 1015 | 73 | |
| GenBank mRNA | BC086967.1 | - | 6 - 7 | 920 | 73 | |
More Information
Set Membership: |
|

fbxo44
Genomic Map