Assay Details
| Gene Symbol: sqor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017881.1 | NP_001017881.1 | 4 - 5 | 506 | 150 | |
| XM_005163161.4 | XP_005163218.1 | 4 - 5 | 716 | 150 | ||
| XM_021470490.1 | XP_021326165.1 | 4 - 5 | 533 | 150 | ||
| GenBank mRNA | BC093431.1 | - | 4 - 5 | 506 | 150 | |
More Information
Set Membership: |
|

sqor
Genomic Map