Assay Details
| Gene Symbol: ppil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034178.1 | NP_001029350.1 | 2 - 3 | 328 | 72 | |
| GenBank mRNA | BC105964.1 | - | 2 - 3 | 328 | 72 | |
More Information
Set Membership: |
|

ppil1
Genomic Map