Assay Details
| Gene Symbol: abcg2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039639.2 | NP_001034728.2 | 5 - 6 | 565 | 138 | |
| XM_005156523.4 | XP_005156580.1 | 5 - 6 | 677 | 138 | ||
| XM_009307089.3 | XP_009305364.1 | 5 - 6 | 746 | 138 | ||
| GenBank mRNA | BC124086.1 | - | 5 - 6 | 565 | 138 | |
| DQ223860.1 | - | 4 - 5 | 510 | 138 | ||
More Information
Set Membership: |
|

abcg2c
Genomic Map