Assay Details
| Gene Symbol: abcg2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039639.2 | NP_001034728.2 | 6 - 7 | 726 | 78 | |
| XM_005156523.4 | XP_005156580.1 | 6 - 7 | 838 | 78 | ||
| XM_009307089.3 | XP_009305364.1 | 6 - 7 | 907 | 78 | ||
| GenBank mRNA | BC124086.1 | - | 6 - 7 | 726 | 78 | |
| BC165778.1 | - | 5 - 6 | 705 | 78 | ||
| DQ223860.1 | - | 5 - 6 | 671 | 78 | ||
More Information
Set Membership: |
|

abcg2c
Genomic Map