Assay Details
| Gene Symbol: abcg2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039639.2 | NP_001034728.2 | 9 - 10 | 1150 | 152 | |
| XM_005156523.4 | XP_005156580.1 | 9 - 10 | 1262 | 152 | ||
| XM_009307089.3 | XP_009305364.1 | 9 - 10 | 1331 | 152 | ||
| GenBank mRNA | BC124086.1 | - | 9 - 10 | 1150 | 152 | |
| BC165778.1 | - | 8 - 9 | 1129 | 152 | ||
| DQ223860.1 | - | 8 - 9 | 1095 | 152 | ||
More Information
Set Membership: |
|

abcg2c
Genomic Map