Assay Details
| Gene Symbol: tiparp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045019.3 | NP_001038484.1 | 5 - 6 | 1652 | 63 | |
| GenBank mRNA | EB973772.1 | - | 1 - 2 | 246 | 63 | |
More Information
Set Membership: |
|

tiparp
Genomic Map