Assay Details
| Gene Symbol: slc27a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077386.1 | NP_001070854.1 | 4 - 5 | 1063 | 66 | |
| GenBank mRNA | BC122420.1 | - | 4 - 5 | 1063 | 66 | |
More Information
Set Membership: |
|

slc27a6
Genomic Map