Assay Details
| Gene Symbol: isoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079981.1 | NP_001073450.1 | 3 - 4 | 604 | 59 | |
| GenBank mRNA | BC124430.1 | - | 3 - 4 | 604 | 59 | |
More Information
Set Membership: |
|

isoc1
Genomic Map