Assay Details
| Gene Symbol: ubfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082830.1 | NP_001076299.1 | 6 - 7 | 770 | 75 | |
| XM_005164210.4 | XP_005164267.1 | 6 - 7 | 854 | 75 | ||
| GenBank mRNA | BC134031.1 | - | 6 - 7 | 770 | 75 | |
More Information
Set Membership: |
|

ubfd1
Genomic Map