Assay Details
| Gene Symbol: vps33a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099973.1 | NP_001093443.1 | 5 - 6 | 629 | 64 | |
| GenBank mRNA | BC095344.1 | - | 5 - 6 | 630 | 64 | |
More Information
Set Membership: |
|

vps33a
Genomic Map