Assay Details
| Gene Symbol: foxa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131299.1 | NP_571374.1 | 1 - 2 | 128 | 90 | |
| GenBank mRNA | AF052245.1 | - | 1 - 2 | 128 | 90 | |
| BC162383.1 | - | 1 - 2 | 120 | 90 | ||
| BC162391.1 | - | 1 - 2 | 120 | 90 | ||
More Information
Set Membership: |
|

foxa3
Genomic Map