Assay Details
| Gene Symbol: pycard | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131495.2 | NP_571570.2 | 2 - 3 | 341 | 73 | |
| GenBank mRNA | BC095025.1 | - | 2 - 3 | 341 | 73 | |
| BC164045.1 | - | 2 - 3 | 332 | 73 | ||
More Information
Set Membership: |
|

pycard
Genomic Map