Assay Details
| Gene Symbol: cxcl14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131627.1 | NP_571702.1 | 2 - 3 | 423 | 77 | |
| GenBank mRNA | AF279919.1 | - | 2 - 3 | 423 | 77 | |
| BC056594.1 | - | 2 - 3 | 330 | 77 | ||
| BC078422.1 | - | 2 - 3 | 359 | 77 | ||
| BC095598.1 | - | 2 - 3 | 260 | 77 | ||
More Information
Set Membership: |
|

cxcl14
Genomic Map