Assay Details
| Gene Symbol: zeb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131709.1 | NP_571784.1 | 6 - 7 | 2400 | 70 | |
| XM_021480009.1 | XP_021335684.1 | 6 - 7 | 4054 | 70 | ||
| GenBank mRNA | AB016799.1 | - | 6 - 7 | 2400 | 70 | |
| BC162144.1 | - | 6 - 7 | 2400 | 70 | ||
More Information
Set Membership: |
|

zeb1b
Genomic Map