Assay Details
| Gene Symbol: lamb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173275.1 | NP_775382.1 | 23 - 24 | 3282 | 75 | |
| GenBank mRNA | AF468049.1 | - | 23 - 24 | 3282 | 75 | |
More Information
Set Membership: |
|

lamb1a
Genomic Map