Assay Details
| Gene Symbol: anxa1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181758.1 | NP_861423.1 | 10 - 11 | 837 | 94 | |
| XM_009301449.1 | XP_009299724.1 | 10 - 11 | 893 | 94 | ||
| GenBank mRNA | AY178793.1 | - | 10 - 11 | 837 | 94 | |
| BC053190.1 | - | 9 - 10 | 820 | 94 | ||
| BC164376.1 | - | 9 - 10 | 818 | 94 | ||
More Information
Set Membership: |
|

anxa1a
Genomic Map