Assay Details
| Gene Symbol: anxa1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181758.1 | NP_861423.1 | 12 - 13 | 1024 | 70 | |
| XM_009301449.1 | XP_009299724.1 | 12 - 13 | 1080 | 70 | ||
| GenBank mRNA | AY178793.1 | - | 12 - 13 | 1024 | 70 | |
| BC053190.1 | - | 11 - 12 | 1007 | 70 | ||
| BC164376.1 | - | 11 - 12 | 1005 | 70 | ||
More Information
Set Membership: |
|

anxa1a
Genomic Map