Assay Details
| Gene Symbol: anxa1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181758.1 | NP_861423.1 | 4 - 5 | 308 | 87 | |
| XM_009301449.1 | XP_009299724.1 | 4 - 5 | 364 | 87 | ||
| GenBank mRNA | AY178793.1 | - | 4 - 5 | 308 | 87 | |
| BC053190.1 | - | 3 - 4 | 291 | 87 | ||
| BC164376.1 | - | 3 - 4 | 289 | 87 | ||
More Information
Set Membership: |
|

anxa1a
Genomic Map