Assay Details
| Gene Symbol: csnk1da | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199583.1 | NP_955877.1 | 4 - 5 | 947 | 71 | |
| XM_005163988.4 | XP_005164045.1 | 4 - 5 | 1021 | 71 | ||
| GenBank mRNA | BC054583.1 | - | 4 - 5 | 947 | 71 | |
| EU127826.1 | - | 4 - 5 | 960 | 71 | ||
More Information
Set Membership: |
|

csnk1da
Genomic Map