Assay Details
| Gene Symbol: camk1ga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199966.1 | NP_956260.1 | 1 - 2 | 121 | 83 | |
| XM_021469304.1 | XP_021324979.1 | 2 - 3 | 202 | 83 | ||
| GenBank mRNA | BC059490.1 | - | 1 - 2 | 121 | 83 | |
More Information
Set Membership: |
|

camk1ga
Genomic Map