Assay Details
| Gene Symbol: camk1ga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199966.1 | NP_956260.1 | 4 - 5 | 462 | 76 | |
| XM_021469304.1 | XP_021324979.1 | 5 - 6 | 543 | 76 | ||
| GenBank mRNA | BC059490.1 | - | 4 - 5 | 462 | 76 | |
More Information
Set Membership: |
|

camk1ga
Genomic Map