Assay Details
| Gene Symbol: klc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200953.2 | NP_957247.1 | 4 - 5 | 417 | 91 | |
| XM_005156633.4 | XP_005156690.1 | 4 - 5 | 905 | 250 | ||
| XM_005156635.4 | XP_005156692.1 | 4 - 5 | 854 | 250 | ||
| XM_005156637.3 | XP_005156694.1 | 4 - 5 | 854 | 250 | ||
| XM_005156638.3 | XP_005156695.1 | 4 - 5 | 854 | 250 | ||
| XM_009307227.3 | XP_009305502.1 | 4 - 5 | 854 | 250 | ||
| GenBank mRNA | BC055591.1 | - | 4 - 5 | 418 | 91 | |
| BC165713.1 | - | 3 - 4 | 377 | 91 | ||
More Information
Set Membership: |
|

klc1a
Genomic Map