Assay Details
| Gene Symbol: cd9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213428.2 | NP_998593.1 | 4 - 5 | 453 | 81 | |
| XM_005164484.2 | XP_005164541.1 | 5 - 6 | 649 | 81 | ||
| GenBank mRNA | BC049036.1 | - | 4 - 5 | 454 | 81 | |
| BC071429.1 | - | 4 - 5 | 431 | 81 | ||
More Information
Set Membership: |
|

cd9b
Genomic Map