Assay Details
| Gene Symbol: cd9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213428.2 | NP_998593.1 | 6 - 7 | 647 | 68 | |
| XM_005164484.2 | XP_005164541.1 | 7 - 8 | 843 | 68 | ||
| GenBank mRNA | BC049036.1 | - | 6 - 7 | 648 | 68 | |
| BC071429.1 | - | 6 - 7 | 625 | 68 | ||
More Information
Set Membership: |
|

cd9b
Genomic Map