Assay Details
| Gene Symbol: tubd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002093.1 | NP_001002093.1 | 5 - 6 | 1129 | 70 | |
| XM_005157418.4 | XP_005157475.1 | 5 - 6 | 924 | 70 | ||
| XM_021481378.1 | XP_021337053.1 | 5 - 6 | 1117 | 70 | ||
| GenBank mRNA | BC071532.1 | - | 5 - 6 | 1129 | 70 | |
More Information
Set Membership: |
|

tubd1
Genomic Map