Assay Details
| Gene Symbol: ndfip1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002503.2 | NP_001002503.1 | 5 - 6 | 584 | 87 | |
| XM_005173872.3 | XP_005173929.1 | 6 - 7 | 621 | 87 | ||
| XM_009295612.2 | XP_009293887.1 | 5 - 6 | 570 | 87 | ||
| GenBank mRNA | BC076296.1 | - | 5 - 6 | 552 | 87 | |
| EH436413.1 | - | 5 - 6 | 593 | 87 | ||
More Information
Set Membership: |
|

ndfip1l
Genomic Map