Assay Details
| Gene Symbol: ndfip1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002503.2 | NP_001002503.1 | 7 - 8 | 762 | 71 | |
| XM_005173872.3 | XP_005173929.1 | 8 - 9 | 799 | 71 | ||
| GenBank mRNA | BC076296.1 | - | 7 - 8 | 730 | 71 | |
| EH436413.1 | - | 7 - 8 | 771 | 71 | ||
More Information
Set Membership: |
|

ndfip1l
Genomic Map