Assay Details
| Gene Symbol: nr2e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003608.1 | NP_001003608.1 | 7 - 8 | 1337 | 80 | |
| GenBank mRNA | BC077146.1 | - | 7 - 8 | 1337 | 80 | |
| EF010862.1 | - | 7 - 8 | 1337 | 80 | ||
More Information
Set Membership: |
|

nr2e1
Genomic Map