Assay Details
| Gene Symbol: picalmb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003741.1 | NP_001003741.1 | 1 - 2 | 458 | 85 | |
| XM_009291635.3 | XP_009289910.1 | 1 - 2 | 603 | 85 | ||
| XM_009291636.3 | XP_009289911.1 | 1 - 2 | 603 | 85 | ||
| XM_009291637.3 | XP_009289912.1 | 1 - 2 | 600 | 85 | ||
| XM_009291638.3 | XP_009289913.1 | 1 - 2 | 605 | 85 | ||
| XM_009291639.3 | XP_009289914.1 | 1 - 2 | 603 | 85 | ||
| XM_009291640.3 | XP_009289915.1 | 1 - 2 | 601 | 85 | ||
| XM_009291641.3 | XP_009289916.1 | 1 - 2 | 600 | 85 | ||
| XM_009291642.3 | XP_009289917.1 | 1 - 2 | 596 | 85 | ||
| XM_009291643.3 | XP_009289918.1 | 1 - 2 | 599 | 85 | ||
| XM_017359233.2 | XP_017214722.1 | 1 - 2 | 606 | 85 | ||
| XM_017359234.2 | XP_017214723.1 | 1 - 2 | 605 | 85 | ||
| XM_017359235.2 | XP_017214724.1 | 1 - 2 | 605 | 85 | ||
| XM_017359236.2 | XP_017214725.1 | 1 - 2 | 603 | 85 | ||
| GenBank mRNA | BC079523.1 | - | 1 - 2 | 458 | 85 | |
More Information
Set Membership: |
|

picalmb
Genomic Map