Assay Details
| Gene Symbol: bsdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004108.2 | NP_001004108.2 | 4 - 5 | 378 | 61 | |
| GenBank mRNA | BC080207.1 | - | 4 - 5 | 385 | 61 | |
| BC155161.1 | - | 4 - 5 | 378 | 61 | ||
More Information
Set Membership: |
|

bsdc1
Genomic Map