Assay Details
| Gene Symbol: phyhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007445.1 | NP_001007446.1 | 10 - 11 | 1098 | 88 | |
| XM_005160976.4 | XP_005161033.1 | 11 - 12 | 993 | 88 | ||
| XM_009295122.3 | XP_009293397.1 | 11 - 12 | 957 | 88 | ||
| GenBank mRNA | BC085396.1 | - | 10 - 11 | 1098 | 88 | |
More Information
Set Membership: |
|

phyhd1
Genomic Map